Polypropylene Stirring Rods
6 mm, 150 mm
Catalog # 470310-636
Supplier: EISCO SCIENTIFIC LLC
Supplier #: CH0757B
Jump to:
Product Details & Documents
Perfect for mixing liquids and chemicals in the laboratory.
Specifications
- Ø:6 mm
- Length:150 mm