$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Mexico
ContactUsLinkComponent
CABLE USB A-MINIDIN 8P PLUG
85014-180
:
MAS INSTRUMENTOS, S.A. DE C.V. MX
CABLE USB A-MINIDIN 8P PLUG
85014-180
:
MAS INSTRUMENTOS, S.A. DE C.V. MX
:
62151000
Frequently Bought Together
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH
VWR® Light-Duty Tissue Wipers
: VWR International
VWR® Disposable Petri Dishes
: VWR International
VWR® Nitrile Examination Gloves, Blue
: VWR International
VWR® Universal Pipette Tips
: VWR International
VWR® Microcentrifuge Tubes, Polypropylene
: VWR International