Differential Counters
5-Key Differential, 9.75L×3.25W×2"H
Catalog # 470309-552
Supplier: LW SCIENTIFIC SE
Número de proveedor #: CTL-DIFM-05KY
Jump to:
Product Details & Documents
Gear and lever reliability and feel.
- Lightweight and portable
- Right and left reset dials
- Non-slip bottom
- Bell alarm for 100th cell counted
Traditionally, differential counters have been manual "gear and lever" machines, and many lab techs are accustomed to the feel of the levered keys. We offer both 5-key and 8-key manual counters for those traditional techs to count Baso, Eosin, Myelo, Juven, Stab, Seg, Lymph, and Mono cells. These counters feature pictures and names of each cell for easier identification. These manual counters are economical, durable, and will provide years of use, even in the most demanding laboratory settings.
Especificaciones
- Description:5-Key Differential
- Dimensions:9.75L×3.25W×2"H