Orders LinkContactUsLinkComponent
Turbidity Standards, LaMotte
Turbidity Standards, LaMotte
1.0 FNU
Catalog # 470308-898
Supplier #:  1453
Turbidity Standards, LaMotte
Catalog # 470308-898
CAS Number:  
Turbidity Standards, LaMotte
Turbidity Standards, LaMotte
1.0 FNU
CAS Number:  
Price
$2,248.21
Log in for availability
Each (60ml)
Product Details & Documents
LaMotte Turbidity Standards 10.0 NTU (2020WE) are essential calibration solutions designed for precise turbidity measurement in water testing. These standards ensure accurate and reliable calibration for turbidity meters, supporting water quality professionals and educators in maintaining optimal water clarity testing.
  • Accurate calibration – Provides a stable and precise 10.0 NTU turbidity reference standard, ensuring your turbidity meter delivers accurate readings
  • Consistent results – Designed to maintain consistency, ensuring repeatable and reliable results for environmental monitoring, water treatment, and laboratory testing
  • Easy integration – Compatible with LaMotte turbidity meters and other water testing equipment, streamlining your calibration process
  • Durable and long-lasting – Comes with a sealed bottle to prevent contamination, ensuring that the turbidity standards remain intact over time
  • Essential for water quality monitoring – Perfect for professionals in water quality testing, industrial applications, and environmental science labs
LaMotte Turbidity Standards 10.0 NTU (2020WE) are essential for calibrating turbidity meters used in water quality testing. These standards ensure accurate, reliable, and consistent results for environmental monitoring, laboratory experiments, and industrial applications.
The 10.0 NTU standard is easy to use and integrates seamlessly with LaMotte turbidity meters, making it ideal for professionals and educators alike. It helps maintain precise measurements for water clarity, crucial for accurate data in water treatment and scientific studies.
Reliable and simple to use, these turbidity standards are a must-have for accurate water quality analysis.
Documents
Specifications
Specifications
  • Turbidity:
    1.0 FNU
Specifications