Ward's® Kick Net
Kick Net Set, 115 cm, 122 cm
470308-238
: B11235H
For drag style collecting.
- Durable netting
- Easily stored and dried
- Simple assembly in field
- Mesh size is 200 microns
- Aligned with GLOBE protocols
The Ward's Kick Net is everything you could want in a student kick net. The square meter collecting area is large enough to collect sufficient samples, but small enough for a single student to manage if necessary, although two students are best. Each end is stabilized by a large pine pole that won’t collapse in a current. The base and sides of the net are framed in muslin sleeves, giving god support and net control.
- Width:122 cm
- Description:Kick Net Set
- Length:115 cm