Order Entry
Mexico
Orders LinkContactUsLinkComponent
 

Ward's® Kick Net

Kick Net, 115 cm, 109 cm
Catalog # 470308-226
Supplier:   SHIVDIAL SUD & SONS
 :  B11235H
Ward's® Kick Net
Catalog # 470308-226
Ward's® Kick Net
Ward's® Kick Net

Kick Net, 115 cm, 109 cm

Price
$497.67
Log in Para disponibilidad
Each (1UN) Case de 50 (1UN)
Product Details & Documents

For drag style collecting.

  • Durable netting
  • Easily stored and dried
  • Simple assembly in field
  • Mesh size is 200 microns
  • Aligned with GLOBE protocols

The Ward's Kick Net is everything you could want in a student kick net. The square meter collecting area is large enough to collect sufficient samples, but small enough for a single student to manage if necessary, although two students are best. Each end is stabilized by a large pine pole that won’t collapse in a current. The base and sides of the net are framed in muslin sleeves, giving god support and net control.

Especificaciones
  • Width:
    109 cm
  • Description:
    Kick Net
  • Length:
    115 cm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.