Orders LinkContactUsLinkComponent
Ward's® Manual Pipette Controller
Ward's® Manual Pipette Controller
Manual Pipette Controller
Catalog # 470307-996
Supplier:   MTC BIO, INC.
Supplier #:  P6420_WARD
Ward's® Manual Pipette Controller
Catalog # 470307-996
Supplier:  MTC BIO, INC.
Ward's® Manual Pipette Controller
Ward's® Manual Pipette Controller
Manual Pipette Controller
Price
$1,835.67
Log in for availability
Each (1UN)
Product Details & Documents
Ward's® Manual pipette controller is the precise and easy to use manual pipette controller that's always ready to use.
  • For glass or plastic pipettes
  • Accommodates pipettes up to 100 ml
  • Easy, single lever control
  • Autoclavable lower body
  • Fingertip speed control
After attaching a pipette, one squeeze of its large rubber filling bulb is all it takes to start pipetting. There are no wires, no tubing, and no battery to recharge.
Begin aspirating by touching the EZ-Flow™ lever which is designed to precisely control the filling speed, based on how gently or firmly the lever is pressed.
A built-in replaceable filter ensures optimum flow rate and protects the controller from liquid penetration and overfill. A separate button provides a positive blow-out function when required.
Specifications
  • Description:
    Manual Pipette Controller