Order Entry
Mexico
Orders LinkContactUsLinkComponent
 

Ebonite Friction Rod, Eisco Scientific

Friction Rod, Ebonite
  470307-036
 :  PH0856A
Ebonite Friction Rod, Eisco Scientific
  470307-036
Ebonite Friction Rod, Eisco Scientific
Ebonite Friction Rod, Eisco Scientific

Friction Rod, Ebonite

 
$131.67
   
Each (1UN)
 

Used to study static and friction.

  • Length: 30 cm
  • Diameter: 12 mm
 
  • Description:
    Friction Rod, Ebonite