Orders LinkContactUsLinkComponent
biOrb® Easy Plant Sets
biOrb® Easy Plant Sets
Kelp Set, Pink, Medium
Catalog # 470234-963
Supplier:   GEOGLOBAL PARTNERS
Supplier #:  46082
biOrb® Easy Plant Sets
Catalog # 470234-963
Supplier:  GEOGLOBAL PARTNERS
biOrb® Easy Plant Sets
biOrb® Easy Plant Sets
Kelp Set, Pink, Medium
Price
$510.05
Log in for availability
Paquete de 2 Case de 4 (2UN)
Product Details & Documents
Quickly add beauty to your aquaria.
  • Artificial plants
  • Suitable for all aquaria
  • Weighted with ceramic ball to prevent floating and tipping
The biOrb® Easy plant packs are a range of plastic plants designed for use in the biOrb®. Instead of a large plastic base these plants have a smooth weighted ball which is easier to fit into the ceramic media in the biOrb® and holds it in place.
The large pack is suitable for 16 gallon/60 l and larger biOrbs. The height of the tallest plant in the twin pack is 15.35"/39 cm, and the base width is 1.57"/4 cm. The medium pack is suitable for 8 gallon/30 l and larger biOrbs. The height of the tallest plant in the twin pack is 11.42"/29 cm, and the base width is 1.5"/3.8 cm. The small pack is suitable for all biOrbs. The height of the tallest plant in the twin pack is 7.87"/20 cm, and the base width is 1.38"/3.5 cm.
Delivery information: Each set includes two plants.
Specifications
  • Color:
    Pink
  • Description:
    Kelp Set
  • Size:
    Medium