Orders LinkContactUsLinkComponent
Tapes, Budnick Converting
Tapes, Budnick Converting
VHB Double Coated Acrylic Foam Tape, White, 36 Yards, 0.165", 15 mil.
Catalog # 76246-602
Supplier #:  4920M165W36LWHT
Tapes, Budnick Converting
Catalog # 76246-602
Tapes, Budnick Converting
VHB Double Coated Acrylic Foam Tape, White, 36 Yards, 0.165", 15 mil.
Price
$1,066.02
Log in for availability
Each (1UN)
Product Details & Documents
Documents
Specifications
  • Width:
    0.165"
  • Color:
    White
  • Description:
    VHB Double Coated Acrylic Foam Tape
  • Length:
    36 Yards
  • Thickness:
    15 mil.