Orders LinkContactUsLinkComponent
Tapes, Budnick Converting
Tapes, Budnick Converting
Kapton P221 Polyimide Film Tape with Silicone Adhesive, Amber, 36 Yards, 0.375", —
Catalog # 76246-118
Supplier #:  P221P375W36L
Tapes, Budnick Converting
Catalog # 76246-118
Tapes, Budnick Converting
Kapton P221 Polyimide Film Tape with Silicone Adhesive, Amber, 36 Yards, 0.375", —
Price
$15.10
Log in for availability
Each (1UN) Case de 64 (1UN)
Product Details & Documents
Documents
Specifications
  • Width:
    0.375"
  • Color:
    Amber
  • Description:
    Kapton P221 Polyimide Film Tape with Silicone Adhesive
  • Length:
    36 Yards
  • Thickness: