Tapes, Budnick Converting
Polyester Film Tape, Black, 72 Yards, 0.25", —
Catalog # 76242-716
Supplier: BUDNICK CONVERTING INC
Número de proveedor #: 850M250W72LBLK
Jump to:
Product Details & Documents
Documents
Certificates
Search by Lot NumberSpecifications
- Width:0.25"
- Color:Black
- Description:Polyester Film Tape
- Length:72 Yards
- Thickness:—
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



