Orders LinkContactUsLinkComponent
SnapLok™ Adapter Assemblies, Nordson EFD
SnapLok™ Adapter Assemblies, Nordson EFD
Syringe Barrel Adapter Assembly, Acetal, 3' Hose, 30/55 cc, Yellow
Catalog # 76237-762
Número de proveedor #:  7005011
SnapLok™ Adapter Assemblies, Nordson EFD
Catalog # 76237-762
SnapLok™ Adapter Assemblies, Nordson EFD
Syringe Barrel Adapter Assembly, Acetal, 3' Hose, 30/55 cc, Yellow
Price
$1,530.97
Log in Para disponibilidad
Each (1UN)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
One-piece molded SnapLok™ adapter provides safe attachment of syringe barrel.
Delivery information: Includes 5/32" OØ flexible polyurethane tubing, Buna N O-ring, quick-connect and safety clip.
Documents
Especificaciones
  • Color:
    Yellow
  • Description:
    Syringe Barrel Adapter Assembly, Acetal, 3' Hose, 30/55 cc
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 