Order Entry
Mexico
Orders LinkContactUsLinkComponent
ACETONE AR 4L

ACETONE AR 4L

Catalog # 85007-024
Número de proveedor #:  9006-03
ACETONE AR 4L
Catalog # 85007-024
CAS Number:  
ACETONE AR 4L
CAS Number:  
Price
$9,650.37
Log in Para disponibilidad
Each (4l)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.