Orders LinkContactUsLinkComponent
Safety Supply Kit
Safety Supply Kit
L
Catalog # 470303-214
Número de proveedor #:  DR20181200-L
Safety Supply Kit
Catalog # 470303-214
Safety Supply Kit
Safety Supply Kit
L
Price
$1,047.49
Log in Para disponibilidad
Each (1UN) Case de 5 (1UN)
Product Details & Documents
Perfect all-in-one safety kit for students.
  • Various lab coat sizes available
Each student should be equipped with one of these kits before beginning any laboratory course. Sizing for gloves and laboratory coats are as follows. XX-small lab coats to small lab coats will receive a pack of small nitrile gloves. Medium lab coats will receive medium nitrile gloves. Large lab coats will receive large nitrile gloves, and X-large to 3X-large lab coats will receive X-large nitrile gloves.
Delivery information: This Safety Kit includes a sized laboratory coat, 10 pack of sized nitrile gloves and a pair of indirect vent splash goggles.
Especificaciones
  • Clothing Size:
    L
  • Cat. No.:
    470303-214
  • Sleeve or leg length:
    Long
  • Garment application:
    Laboratory
  • Gender:
    Unisex
  • Closure type:
    Buttons