Skip to Header
Skip to Main Content
Skip to Footer
Avanzado
Avanzado
SDS
Certificados
Estructura química
Datos del fabricante
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Mexico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
MATRAZ VOL CLARO HD CLASE 10ML
Catalog # 85005-970
Supplier:
DWK LIFE SCIENCES S. DE R.L. DE MX
Certificates
Expand image
MATRAZ VOL CLARO HD CLASE 10ML
Catalog # 85005-970
Supplier:
DWK LIFE SCIENCES S. DE R.L. DE MX
Supplier Number:
92820G-10
Certificates
Related Information
Documentation
Certificates
Search by Lot Number
89217-682_4600071735
Documentation
Frequently Bought Together
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Light-Duty Tissue Wipers
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Microcentrifuge Tubes, Polypropylene
Supplier: VWR International
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International