Magnetic Mounting Sets
Magnetic Mount 4
Catalog # 470236-394
Supplier: MARKETLAB, INC.
Número de proveedor #: 13992
Jump to:
Product Details & Documents
Mount a dispenser on any magnetic surface.
- Neoprene-coated magnets protect metal surfaces
- Powerful, industrial-strength 30 lbs. magnets
Easily convert any existing wall mount unit with keyholes for use on metal surfaces with these exclusive magnetic mounting sets.
Especificaciones
- Description:Magnetic Mount 4