Orders LinkContactUsLinkComponent
Magnetic Mounting Sets
Magnetic Mounting Sets
Magnetic Mount 2
Catalog # 470236-392
Supplier:   MARKETLAB, INC.
Número de proveedor #:  13991
Magnetic Mounting Sets
Catalog # 470236-392
Supplier:  MARKETLAB, INC.
Magnetic Mounting Sets
Magnetic Mounting Sets
Magnetic Mount 2
Price
$3,839.79
Log in Para disponibilidad
Each (2UN) Paquete de 2
Product Details & Documents
Mount a dispenser on any magnetic surface.
  • Neoprene-coated magnets protect metal surfaces
  • Powerful, industrial-strength 30 lbs. magnets
Easily convert any existing wall mount unit with keyholes for use on metal surfaces with these exclusive magnetic mounting sets.
Especificaciones
  • Description:
    Magnetic Mount 2