Orders LinkContactUsLinkComponent
Crazy Circuits Sets
Crazy Circuits Sets
Crazy circuits starter set
Catalog # 470234-670
Supplier:   BROWN DOG GADGETS
Número de proveedor #:  K1016
Crazy Circuits Sets
Catalog # 470234-670
Supplier:  BROWN DOG GADGETS
Crazy Circuits Sets
Crazy Circuits Sets
Crazy circuits starter set
Liquidación
$43.00
Oferta
Válido hasta 31/12/2026
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
Create your own brick-based electronics projects in minutes with these Crazy Circuits sets.
  • Design your own projects from scratch, or follow one of our many project guides
  • Compatible with LEGO™ and similar brick building systems
  • No soldering or special tools required
Use the included brick baseplates, or add Crazy Circuits parts to any LEGO project to make it light up! These kits come 100% ready to go out of the box, which means no soldering or special equipment needed. They cover topics such as circuitry, programming, logic, design, art, and design. Detailed directions, project guides, educational resources, and videos can be found online.
The Makerspace set has everything you need to get an entire classroom creating projects with Crazy Circuits. Use the bit boards and robotics boards to control servo motors and add functionality. Create wearable technology using the conductive thread.
 : Starter set includes:
(10) Jumbo 10 mm LEDs (red, amber, green, blue, white); (5) CR2032 batteries; (2) coin cell battery holders; (2) pushbutton switches; (1) slide switch; (1) roll of conductive maker tape – ¹/₈"×5 m; (1) brick baseplate (8×16 studs); (1) plastic organizer; instructional cards.
Makerspace set includes:
(65) LEDs (red, amber, green, blue, white); (50) CR2032 batteries; (10) coin cell battery holders; (15) switches; (10) screw terminals; (10) brick baseplates (8×16 studs); (10) rolls of conductive maker tape – ¹/₈"×5 m; (8) LEGO-compatible servo motors and connector pieces; (6) USB cables; (5) NeoPixel LED strips; (5) ribbon cables; (5) piezoelectric speakers; (5) potentiometers; (5) 2AAA battery holders; (5) spools conductive thread; (2) bundles of 5 alligator clip jumper wires; (2) bit boards; (2) Micro:bits; (2) plastic organizers; instructional cards.
Especificaciones
  • Description:
    Crazy circuits starter set
Accessories
Información del producto
  • Catalog #
    Availability
    Price
  • Conductive maker tape, ⅛" × 5 m
    Conductive maker tape, ⅛" × 5 m
    470234-664
    Log in Para disponibilidad
    Each (1UN) - $180.00