Disposable Micropipet Tips
1000 - 5000 µl, 100 Tips/Bag
Catalog # 470233-654
Supplier: EDVOTEK, INC.
Número de proveedor #: 637-2
Jump to:
Product Details & Documents
High quality micropipette tips to meet any lab's needs.
- Certified DNase, RNase and pyrogen free
- Material: PP
- Autoclavable
- Designed to fit most standard micropipettes
All quantities are supplied in bulk bags.
Especificaciones
- Volume:1000 - 5000 µl
- Packed:100 Tips/Bag