Orders LinkContactUsLinkComponent
Science Take-Out® Brittle Bones: A Density Problem
Science Take-Out® Brittle Bones: A Density Problem
Brittle bones, unassembled 10 pack, 10
Catalog # 470232-166
Número de proveedor #:  STO-154U
Science Take-Out® Brittle Bones: A Density Problem
Catalog # 470232-166
Science Take-Out® Brittle Bones: A Density Problem
Science Take-Out® Brittle Bones: A Density Problem
Brittle bones, unassembled 10 pack, 10
Price
$3,350.00
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
What is Osteoporosis?
  • Available as a single kit with material for one student or group
  • Unassembled 10 pack for additional savings
Three generations of a family learn how osteoporosis (low bone density) results in bones that are brittle and easily broken. Students will design and assemble models of bones to represent bone densities at different ages to determine their densities. This activity also helps students learn to interpret models, graphs, news articles, and posters to relate bone density, osteoporosis, and bone fracture risk.
Documents
Especificaciones
  • Description:
    Brittle bones, unassembled 10 pack
  • No. of Lab Groups:
    10