Ward’s® Adjustable Volume Micropipettes
1000 μL Micropipette, 100 - 1000 µl, 5.0 µl
Catalog # 470231-602
Supplier: DLAB SCIENTIFIC INC.
: 7010301014
Jump to:
Product Details & Documents
The Wards® Micropipette line is fully autoclavable, helping to prevent cross contamination. The ergonomic design is ambidextrous, providing all users with an excellent operating experience.
- Fully autoclavable
- Slim, lightweight ergonomic design
- Compatible with most universal tip brands
- Tool included for quick and easy recalibration
- Purchase individually, or as one of our available sets
Personal Pipette Set comes with one of each pipette shown, plus a storage carousel.
Classroom Pipette Set comes with eight of each pipette shown, plus eight storage carousels.
Especificaciones
- Volume:100 - 1000 µl
- Description:1000 μL Micropipette
- Increments:5.0 µl