Orders LinkContactUsLinkComponent
Ward’s® Adjustable Volume Micropipettes
Ward’s® Adjustable Volume Micropipettes
200 μL Micropipette, 20 - 200 µl, 1.0 µl
Catalog # 470231-600
Número de proveedor #:  7010301009
Ward’s® Adjustable Volume Micropipettes
Catalog # 470231-600
Ward’s® Adjustable Volume Micropipettes
Ward’s® Adjustable Volume Micropipettes
200 μL Micropipette, 20 - 200 µl, 1.0 µl
Price
$1,606.62
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
The Wards® Micropipette line is fully autoclavable, helping to prevent cross contamination. The ergonomic design is ambidextrous, providing all users with an excellent operating experience.
  • Fully autoclavable
  • Slim, lightweight ergonomic design
  • Compatible with most universal tip brands
  • Tool included for quick and easy recalibration
  • Purchase individually, or as one of our available sets
Personal Pipette Set comes with one of each pipette shown, plus a storage carousel.
Classroom Pipette Set comes with eight of each pipette shown, plus eight storage carousels.
Especificaciones
  • Volume:
    20 - 200 µl
  • Description:
    200 μL Micropipette
  • Increments:
    1.0 µl