Accessories for Solvent Pump, Bürkle
Thread Adapter, 38 mm to 2" BSP Inner, Used to change a 38 mm thread of a canister to a 2" BSP thread; Suitable for US tin foil canisters with an opening of 38 mm
76121-370
: 0600-2018
- Description:Thread Adapter, 38 mm to 2" BSP Inner
- For Use With:Used to change a 38 mm thread of a canister to a 2" BSP thread; Suitable for US tin foil canisters with an opening of 38 mm