Orders LinkContactUsLinkComponent
Accessories for Solvent Pump, Bürkle
Thread Adapter, 38 mm to 2" BSP Inner, Used to change a 38 mm thread of a canister to a 2" BSP thread; Suitable for US tin foil canisters with an opening of 38 mm
  76121-370
 :   BURKLE INC
 :  0600-2018
Accessories for Solvent Pump, Bürkle
  76121-370
Accessories for Solvent Pump, Bürkle
Accessories for Solvent Pump, Bürkle
Thread Adapter, 38 mm to 2" BSP Inner, Used to change a 38 mm thread of a canister to a 2" BSP thread; Suitable for US tin foil canisters with an opening of 38 mm
$3,007.96
Each (1UN)
  • Description:
    Thread Adapter, 38 mm to 2" BSP Inner
  • For Use With:
    Used to change a 38 mm thread of a canister to a 2" BSP thread; Suitable for US tin foil canisters with an opening of 38 mm