$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Mexico
ContactUsLinkComponent
OREJERAS 27 DECIBELES 3M
85003-376
:
GRAINGER,S.A. DE C.V. MX
OREJERAS 27 DECIBELES 3M
85003-376
:
GRAINGER,S.A. DE C.V. MX
:
MXX4A
Frequently Bought Together
VWR® Disposable Petri Dishes
: VWR International
VWR® Nitrile Examination Gloves, Blue
: VWR International
VWR® Light-Duty Tissue Wipers
: VWR International
VWR® Universal Pipette Tips
: VWR International
VWR® Microcentrifuge Tubes, Polypropylene
: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH