Orders LinkContactUsLinkComponent
DR-MED® Laparoscopic Training Instruments
Laparoscopic forceps
  470231-228
 :  DR786922
DR-MED® Laparoscopic Training Instruments
  470231-228
DR-MED® Laparoscopic Training Instruments
DR-MED® Laparoscopic Training Instruments
Laparoscopic forceps
$3,861.63
Each (1UN)
Reasonably priced practice tools.
  • The same tools you would use in surgery
  • 360 degree rotation
  • 19" length
Learn skills using tools with the same features and grips as the surgical instruments you will be using. Securely hold and manipulate tissues with the grasper and perform fine manipulations with the forceps. Practice removing tissue with the scissors, and closing internally with the needle driver.
 : For Training Purposes Only. Not for human or animal use.
  • Description:
    Laparoscopic forceps