DR-MED® Laparoscopic Training Modules
Suture Module
Catalog # 470231-222
Supplier: DR INSTRUMENTS INC. SE
Número de proveedor #: DR786932
Jump to:
Product Details & Documents
Pre-assembled exercises.
- Build skills using laparoscopic tools
- Useable with any trainer
Use these laparoscopic training modules to learn the control necessary for surgery. Use the clamp module to practice the fine control needed to transfer small materials across a surgical field. Each bead can be moved to a post and back. The perforate module gives you practice using both hands to thread and unthread a pipe cleaner through a series of eye screws. Once you think you have it mastered, you can rotate the eyelets and create a new, more difficult, path. Finally practice sutures using the suture module. Seven independent areas allow you to practice on curved, straight, variable and circular sections.
Especificaciones
- Description:Suture Module