Eisco® Electrodes
Brass electrode, strip, 100×19 mm
470230-332
: PH0927B
Individual metal electrodes for electrochemical experiments.
Each electrode is stamped with its metal type for easy identification
- Durable and reusable for multiple lab sessions
Eisco electrodes are an essential tool for students exploring electrochemical reactions and principles. Each electrode strip measures approximately 100×19 mm (4×0.75"), is crafted from high-quality metals and clearly stamped for easy identification.These electrodes are perfect for hands-on learning in topics such as oxidation-reduction reactions, electrode potentials, and galvanic cells.
- Description:Brass electrode, strip
- Size:100×19 mm