Friction Rods
Ebonite Rod
470230-306
: PH0856A
For your Electrostatic Needs!
- Excellent quality
Friction rods are used for producing static charges when rubbed against materials such as wool, flannel, silk, and fur.
- Description:Ebonite Rod
For your Electrostatic Needs!
Friction rods are used for producing static charges when rubbed against materials such as wool, flannel, silk, and fur.