Orders LinkContactUsLinkComponent
Neodymium Disk Magnet
Neodymium Disk Magnet, 0.5" Diameter, 0.125 " Thick
Catalog # 470229-350
Neodymium Disk Magnet
Catalog # 470229-350
Supplier:  
Neodymium Disk Magnet
Neodymium Disk Magnet
Neodymium Disk Magnet, 0.5" Diameter, 0.125 " Thick
Discontinued
El producto ha sido descontinuado. Sin embargo, encontramos productos alternativos similares.
Alternativas
Product Details & Documents
A powerful magnet for a variety of experiments.
  • Very high resistance to demagnetization
  • High energy for size
  • Good in ambient temperature
Neodymium Disk Magnets (Nd-Fe-B) are made from the most powerful magnetic material in the world. They boast extreme strength in a small size. All of the magnets have a grade of 35 with a Gauss rating of 12,200. Maximum operating temperature is 80°C (176°F).
Especificaciones
  • Description:
    Neodymium Disk Magnet, 0.5" Diameter, 0.125 " Thick