Crystallizing Dish
2000 ml, 190 mm, 100 mm
Catalog # 470229-206
Supplier: EISCO SCIENTIFIC LLC
Número de proveedor #: CH0066C
Jump to:
Product Details & Documents
Ideal for storage and crystallization.
- Borosilicate glass
- Flat bottom
- Easy pour spout
Made from high quality borosilicate glass, these crystallizing dishes are perfect for use in any laboratory. Dishes feature flat bottoms and sides, with an easy pour spout. Available in five sizes.
Especificaciones
- Ø:190 mm
- Capacity:2000 ml
- Height:100 mm