Orders LinkContactUsLinkComponent
Resonance Apparatus
Resonance Apparatus
Replacement Cup
Catalog # 470229-058
Número de proveedor #:  PH0720CUP
Resonance Apparatus
Catalog # 470229-058
Resonance Apparatus
Resonance Apparatus
Replacement Cup
Price
$421.07
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
With the Resonance Apparatus, students investigate the effect of frequency on wavelength.
  • Durable
  • Inexpensive
  • For Determining the Wavelength and Velocity of Sound
  • Weight: 4.8 lb.
  • Height: 39" after Assembly
An 98cm, transparent, graduated plastic tube is connected to a water reservoir. Both the tube and the water reservoir are securely mounted to a metal support. The water reservoir may be raised or lowered to adjust the air column.
The apparatus weighs 4.8lbs. and is 39" when assembled. The tube is 2.5cm dia. A tuning fork is required.
Especificaciones
  • Description:
    Replacement Cup