Orders LinkContactUsLinkComponent
Graduated Reagent Bottle
Graduated Reagent Bottle
100 ml, Amber
Catalog # 470228-984
Número de proveedor #:  CH0165A
Graduated Reagent Bottle
Catalog # 470228-984
Graduated Reagent Bottle
Graduated Reagent Bottle
100 ml, Amber
Price
$371.49
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
Mess-free pouring.
  • Screw cap
  • Borosilicate glass
  • Poring ring
  • Both cap and bottle are autoclavable
These economy graduated reagent bottles are the perfect addition to any laboratory. Made from borosilicate glass, these bottles have a convienent screw cap and a drip-free pouring ring to help maintain a clean work environment.
Especificaciones
  • Capacity:
    100 ml
  • Color:
    Amber