Graduated Reagent Bottle
100 ml, Clear
Catalog # 470228-974
Supplier: EISCO SCIENTIFIC LLC
Supplier #: CH0164A
Jump to:
Product Details & Documents
Mess-free pouring.
- Screw cap
- Borosilicate glass
- Poring ring
- Both cap and bottle are autoclavable
These economy graduated reagent bottles are the perfect addition to any laboratory. Made from borosilicate glass, these bottles have a convienent screw cap and a drip-free pouring ring to help maintain a clean work environment.
Documents
Specifications
- Capacity:100 ml
- Color:Clear