Order Entry
Mexico
Orders LinkContactUsLinkComponent
Graduated Cylinders

Graduated Cylinders

500 ml, 5.0 ml, ±2.00 ml
Catalog # 470228-970
Número de proveedor #:  CH0341FBL
Graduated Cylinders
Catalog # 470228-970
Graduated Cylinders
Graduated Cylinders

500 ml, 5.0 ml, ±2.00 ml

Price
$746.74
Log in Para disponibilidad
Each (1UN)
Product Details & Documents

Class A Cylinders.

  • Blue graduations
  • Hexagonal base
  • Plastic cylinder guard

These Class A Graduated Cylinders are the perfect laboratory companion. Made from high quality borosilicate glass with a hexagonal base. Supplied complete with protective cylinder guard. Cylinders comply with ASTM E1272 standard.

Especificaciones
  • Graduation Interval:
    5.0 ml
  • Capacity:
    500 ml
  • Tolerance:
    ±2.00 ml
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.