Volumetric Flasks
250 ml, 16, ±0.120 ml
Catalog # 470228-956
Supplier: EISCO SCIENTIFIC LLC
Número de proveedor #: CH0441E
Jump to:
Product Details & Documents
Class A Flasks.
- Blue graduations
- Glass stopper
Perfect for everyday use in the laboratory, these borosilicate glass flasks are marked with blue graduations for easy reading and fitted with a solid glass stopper. In compliance with ASTM-E288 standard.
Especificaciones
- Capacity:250 ml
- Stopper No.:16
- Tolerance:±0.120 ml