Volumetric Flasks
100 ml, 13, ±0.080 ml
Catalog # 470228-954
Supplier: EISCO SCIENTIFIC LLC
: CH0441C
Jump to:
Product Details & Documents
Class A Flasks.
- Blue graduations
- Glass stopper
Perfect for everyday use in the laboratory, these borosilicate glass flasks are marked with blue graduations for easy reading and fitted with a solid glass stopper. In compliance with ASTM-E288 standard.
Especificaciones
- Capacity:100 ml
- Stopper No.:13
- Tolerance:±0.080 ml