Volumetric Flasks
25 ml, 9, ±0.030 ml
Catalog # 470228-950
Supplier: EISCO SCIENTIFIC LLC
: CH0441A
Jump to:
Product Details & Documents
Class A Flasks.
- Blue graduations
- Glass stopper
Perfect for everyday use in the laboratory, these borosilicate glass flasks are marked with blue graduations for easy reading and fitted with a solid glass stopper. In compliance with ASTM-E288 standard.
Especificaciones
- Capacity:25 ml
- Stopper No.:9
- Tolerance:±0.030 ml