Polystyrene Storage Jars
8 oz., 89 mm
Catalog # 470227-552
Supplier: PLASTIC BOTTLES INC.
Supplier #: 181634
Jump to:
Product Details & Documents
Ideal for specimen storage.
- Clear polystyrene
- Wide-mouth
- Aligned with GLOBE protocols
Ordering information: Metal jar caps sold separately.
Specifications
- Cap Size:89 mm
- Capacity:8 oz.