Orders LinkContactUsLinkComponent
Iron Meteorite
Iron Meteorite
Iron Meteorite, ¾ - 1"
Catalog # 470226-698
Supplier:   Avantor
Iron Meteorite
Catalog # 470226-698
Supplier:  Avantor
Iron Meteorite
Iron Meteorite
Iron Meteorite, ¾ - 1"
Price
$220.20
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
Fragments in natural condition, many showing concave forms and fusion crusts. Sizes are approximate.
  • Available in two sizes
  • Slicked in oil to prevent rusting
  • Protected in bubble wrap and a sealed bag
Especificaciones
  • Description:
    Iron Meteorite
  • Specimen Dimensions:
    ¾ - 1"