VWR® Sharps Container Systems, Red
1 qt., Stackable, 8.9W×8.9D×17.8H cm (3.5W×3.5D×7"H), Yellow
Sharps container systems provide solutions for the safe and effective containment and disposal of contaminated needles, syringes, and other sharps. Choose from three different systems in a variety of sizes.
- Puncture-resistant
- Autoclave-safe and incinerator-safe
- Transparent, locking lids
- Needle removal ports
The extended neck sharps containers feature a funneled vertical drop and extended handles. Puncture-resistant and autoclave- and incinerator-safe.
- Capacity:1 qt.
- Color:Yellow
- Dimensions:8.9W×8.9D×17.8H cm (3.5W×3.5D×7"H)
- Style:Stackable



