Knife Switches
Knife Switch Double Pole, Double Throw
Catalog # 470225-208
Supplier: VWR International
Jump to:
Product Details & Documents
Bigger design for easy connections.
- Easy-to-connect
- Unbreakable
- Teaching tool for open circuit lessons
- Rated at 5A
- Four configurations available
Basic components for any electrical lab, these switches are anchored onto plastic bases that allow visible connections for easy use. Educators use Plastic Knife Switches to explain open circuits
Specifications
- Description:Knife Switch Double Pole, Double Throw