Orders LinkContactUsLinkComponent
Knife Switches
Knife Switches
Knife Switch Double Pole, Double Throw
Catalog # 470225-208
Supplier:   VWR International
Knife Switches
Catalog # 470225-208
Supplier:  VWR International
Knife Switches
Knife Switches
Knife Switch Double Pole, Double Throw
Clearance
$2.60
Sale
Valid until 31/12/2026
Log in for availability
Each (1UN) Case de 50 (1UN)
Product Details & Documents
Bigger design for easy connections.
  • Easy-to-connect
  • Unbreakable
  • Teaching tool for open circuit lessons
  • Rated at 5A
  • Four configurations available
Basic components for any electrical lab, these switches are anchored onto plastic bases that allow visible connections for easy use. Educators use Plastic Knife Switches to explain open circuits
Specifications
  • Description:
    Knife Switch Double Pole, Double Throw