Orders LinkContactUsLinkComponent
Knife Switches
Knife Switch Single Pole, Single Throw
  470225-202
Knife Switches
  470225-202
Knife Switches
Knife Switches
Knife Switch Single Pole, Single Throw
$47.83
Each (1UN) Case de 50 (1UN)
Bigger design for easy connections.
  • Easy-to-connect
  • Unbreakable
  • Teaching tool for open circuit lessons
  • Rated at 5A
  • Four configurations available
Basic components for any electrical lab, these switches are anchored onto plastic bases that allow visible connections for easy use. Educators use Plastic Knife Switches to explain open circuits
  • Description:
    Knife Switch Single Pole, Single Throw