SANI-STICK Sampling Kits, Labplas
SANI-STICK kit with wet sponge on handle with buffered peptone water, 600 ml, 140×279 mm (5.5×11")
75849-322
Supplier #: KSS-67310-BPW
The SANI-STICK is designed to collect samples to detect the presence of microbiological contaminations on any surface.
- Allows swabbing of hard-to-reach areas
- Convenient and east to use
- Helps eliminate several time -consuming steps
These sponges on sticks are widely used in the food industry applications such as quality assurance procedures (QA/QC).
Documents
- Capacity:600 ml
- Description:SANI-STICK kit with wet sponge on handle with buffered peptone water
- W×L:140×279 mm (5.5×11")
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



