Orders LinkContactUsLinkComponent
Collapsible Metal
Collapsible Metal Can with Hand Pump and Stopper
  470222-636
 :  PH0201603
Collapsible Metal
  470222-636
Collapsible Metal
Collapsible Metal
Collapsible Metal Can with Hand Pump and Stopper
$1,829.80
Each (1UN)
Dramtically demonstrate the power of atmospheric pressure
  • Demonstrate ideal gas law in a visually spectacular manner
  • Great for classrooms of all sizes
This metal can with stopper and fitting provides an intriguing demonstration of physics principles relating to gas and air pressure. The unit can be purchased including the hand pump and replacement cans are available as well. The can also can be collapsed using boiling water to evacuate air from the can. Includes cap as well as stopper and hose fitting.
  • Description:
    Collapsible Metal Can with Hand Pump and Stopper