Order Entry
Mexico
Orders LinkContactUsLinkComponent
 

Student-Grade Polypropylene Beakers

100 ml (3.4 oz.)
Catalog # 470222-440
Supplier:   SHIVDIAL SUD & SONS
 :  CH10155/3
Student-Grade Polypropylene Beakers
Catalog # 470222-440
Student-Grade Polypropylene Beakers
Student-Grade Polypropylene Beakers

100 ml (3.4 oz.)

Price
$10.77
Log in for availability
Each (1UN) Paquete de 12 (1UN) Case de 48 (1UN)
Product Details & Documents

These simple, stackable, ribbed beakers with raised and permanent graduations are an excellent value.

  • Polypropylene construction
  • Stackable
  • Simple to use
  • Permanent graduations for easy measurements
  • Autoclavable at 121 °C

Perfect for use with acids, bases, and alcohols, they can also be used short-term with hydrocarbons.

Specifications
  • Capacity:
    100 ml (3.4 oz.)