Orders LinkContactUsLinkComponent
Boreal Research Microscope Series
Boreal Research Microscope Series
Binocular Compound Microscope, Semi-Plan
Catalog # 470222-444
Número de proveedor #:  168-SP
Boreal Research Microscope Series
Catalog # 470222-444
Boreal Research Microscope Series
Boreal Research Microscope Series
Binocular Compound Microscope, Semi-Plan
Price
$25,211.26
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
This microscope is ideal for advanced studies and laboratory use.
  • Choice of Objective Lenses: ASC or Semi-Plan
  • LED Illumination
  • Large, Built-In, Low-Profile, Rack-Less Stage
  • Manufacturer's Lifetime Warranty
  • Superb Optical Clarity
  • Color-Coded Objectives
The new Research series incorporates a wide range of features designed for university and laboratory use. Superb optical clarity and reliable mechanical engineering assure years of top performance.
Two grades of objective lenses are available, both of which are parfocal, parcentered, and color coded.
Ordering information: This product includes a manufacturer's warranty.
Especificaciones
  • Description:
    Binocular Compound Microscope, Semi-Plan
  • Cat. No.:
    470222-444
  • Illumination:
    LED Corded
  • Stage:
    Rackless Stage
  • Magnification:
    4X, 10X, 40X, 100X Objectives