Boreal Research Microscope Series
Binocular Compound Microscope, Semi-Plan
Catalog # 470222-444
Supplier: NATIONAL OPTICAL AND SCIENTIFIC SE
Número de proveedor #: 168-SP
Jump to:
Product Details & Documents
This microscope is ideal for advanced studies and laboratory use.
- Choice of Objective Lenses: ASC or Semi-Plan
- LED Illumination
- Large, Built-In, Low-Profile, Rack-Less Stage
- Manufacturer's Lifetime Warranty
- Superb Optical Clarity
- Color-Coded Objectives
The new Research series incorporates a wide range of features designed for university and laboratory use. Superb optical clarity and reliable mechanical engineering assure years of top performance.
Two grades of objective lenses are available, both of which are parfocal, parcentered, and color coded.
Ordering information: This product includes a manufacturer's warranty.
Especificaciones
- Description:Binocular Compound Microscope, Semi-Plan
- Cat. No.:470222-444
- Illumination:LED Corded
- Stage:Rackless Stage
- Magnification:4X, 10X, 40X, 100X Objectives