Orders LinkContactUsLinkComponent
MicroGenesis™ Single Use Microfiber Products, Contec®
MicroGenesis™ Single Use Microfiber Products, Contec®
Replacement MicroGenesis™ Hygienic backer plate, 24/Case, 10.2×40.6 cm (4×16")
Catalog # 75803-204
MicroGenesis™ Single Use Microfiber Products, Contec®
Catalog # 75803-204
Supplier:  
MicroGenesis™ Single Use Microfiber Products, Contec®
MicroGenesis™ Single Use Microfiber Products, Contec®
Replacement MicroGenesis™ Hygienic backer plate, 24/Case, 10.2×40.6 cm (4×16")
Discontinued
Product Details & Documents
MicroGenesis™ Single Use Microfiber Pads feature synthetic construction for the effective removal of both gross contamination and fine particles. Conveniently designed to provide a new pad for each new use, they are compatible with most existing hardware.
  • Compatible with common disinfectants like quats, peroxides, and bleach
  • Simplifies quality control, inventory, and space management, all without cross contamination risk
  • Provides efficient and effective removal of surface contamination
  • Use MicroGenesis™ Floor Pads with MicroGenesis™ Hygienic Backer Plate and appropriate hardware
MicroGenesis™ Microfiber Floor Pads
These 100% synthetic, disposable microfiber floor pads effectively remove fine particles and large debris from floors, providing superior removal of visible contaminants. Designed to be used wet or dry for general facility cleaning in ISO Class 8 environments and above. Compatible with a wide range of disinfectants, these synthetic pads are designed to provide uniform surface coverage. A new pad for each application minimizes cross-contamination risks (both residual and microbial) unlike laundered mops, and does not interfere with disinfectant efficacy like cotton or rayon. Use Contec’s hardware for improved functionality and ergonomic ease.
Especificaciones
  • Description:
    Replacement MicroGenesis™ Hygienic backer plate
  • Size:
    10.2×40.6 cm (4×16")
  • Packaging:
    24/Case