Density Sets
Density Set, Aluminum
Catalog # 470221-106
Supplier: EISCO SCIENTIFIC LLC
Supplier #: PH0113F
Jump to:
Product Details & Documents
Students Learn That the Density of a Substance is Constant, Even as Its Mass and Volume Change.
- Each set has 12 cylindrical bars
- Bars are same diameter but different lengths
- Each set comes in an attractive wooden holder
Specifications
- Description:Density Set, Aluminum