Orders LinkContactUsLinkComponent
Complete Meter Stick Optics Bench
Complete Meter Stick Optics Bench
Meter Stick Optics Bench, No Meter Stick, —
Catalog # 470219-702
Supplier #:  OBSET2-NS
Complete Meter Stick Optics Bench
Catalog # 470219-702
Complete Meter Stick Optics Bench
Complete Meter Stick Optics Bench
Meter Stick Optics Bench, No Meter Stick, —
Price
$207.34
Log in for availability
Each (1UN)
Product Details & Documents
Classical experiment for studying optics on a budget.
  • All components connect to any standard meter stick
Student optical bench contains sufficient equipment for high school physics applications.
Contents:
1 Meter Stick
1 Screen support
2 lens supports for 38mm and 75mm lenses/mirrors
5 Board screens measuring 10 x 125 cm with a millimeter scale on both sides
1 Marker and object
2 Metal supports for meter stick
1 Candle holder
1 Paraffin candle
1 Activity guide
Required but not included are lenses. Focal lengths under 50cm work best.
Specifications
  • Description:
    Meter Stick Optics Bench, No Meter Stick
  • No. of Lab Groups:
Recommendations